Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TGFA Mouse Monoclonal Antibody [clone: 1E8-269]

Supplier: Genetex
GTX11456
Human TGF - alpha. Transforming growth factor-alpha (TGF alpha) is an acid- and heat-stable 50 amino acid protein of 5,500 MW originally discovered in rodents and humans. TGF alpha protien is 33% homologous to epidermal growth factor (EGF). TGF alpha binds to the EGF receptor, mediating tyrosine phosphorylation of the receptor, and promotes anchorage-independent growth of normal rat fibroblasts in soft agar in the presence of transforming growth factor-beta. TGF alpha is secreted by a variety of transformed cells and tumors, embryonic cells and some normal adult cells . TGF alpha bioactivity has also been found in the urine of cancer patients. It has been suggested that it may act as an autocrine growth factor for the induction or maintenance of malignancy.

Recommended Starting Dilutions:ELISA: Max Dilution 1/10Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: TGFA
Clonality: Monoclonal
Clone: 1E8-269
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
1 ml GTX11456 89358-072 Each (1ml) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol TGFA
Antigen Name Transforming Growth Factor, Alpha
Reactivity Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody Type Primary
Clone 1E8-269
Host Mouse
Isotype IgG1
ELISA Yes
Antigen Synonyms TGF-a, ETGF, transforming growth factor alpha, TGF type 1, TGFalpha, EGF-like TGF, TGF-alpha, TGF alpha
Amino Acid Number 32 to 40
Gene ID 7039
Format Tissue culture supernatant
Immunogen Peptide corresponding to amino acid residues 32-40 of human TGF-α linked to thyroglobulin.
Purification Tissue Culture Supernatant containing 0.2M Tris/HCl pH7.4 and 5-10% foetal calf serum
Storage Buffer 0.09% Sodium Azide (NaN3) and 0.1% Bovine Serum Albumin
Storage Temperature Store at +4°C or at -20°C if preferred. This product should be stored undiluted. Storage in frost free freezers is not recommended. Should this product contain a precipitate we recommend microcentrifugation before use.
Tested Applications ELISA

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty