Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Testosterone Mouse Monoclonal Antibody

Supplier: Genetex

GTX26066
89352-162EA 269.01 USD InStock
89352-162
Anti-Testosterone Mouse Monoclonal Antibody
Antibodies
Testosterone is the principal androgen, or male sex hormone. One of the group of compounds known as anabolic steroids, testosterone is secreted by the testes but is also synthesized in small quantities in the ovaries, cortices of the adrenal glands, and placenta, usually from cholesterol. Testosterone is necessary in the fetus for the development of male external genitalia. Testosterone also stimulates protein synthesis and accounts for the greater muscular development of the male. For many years, synthetic steroids similar to testosterone have been used by athletes with the goal of improving performance, although recent medical research has shown that these drugs may have a wide range of harmful side effects.

Type: Primary
Antigen: Testosterone
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mammals

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty