Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TDT Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: HT-1, HT-4, HT-8, HT-9]

Supplier: Genetex

GTX43336
89367-736EA 943.73 USD InStock
89367-736
Anti-TDT Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: HT-1, HT-4, HT-8, HT-9]
Antibodies
Mouse monoclonal antibody [HT-1, HT-4, HT-8, HT-9] to TdT

Type: Primary
Antigen: TDT
Clonality: Monoclonal
Clone: HT-1, HT-4, HT-8, HT-9
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty