Anti-TCR V beta 5 Mouse Monoclonal Antibody

Supplier: GeneTex
GTX21898
89361-108EA 621.82 USD
89361-108
Anti-TCR V beta 5 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: TCR V beta 5
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR