Anti-MAPT Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX24749
89361-478EA 608.78 USD
89361-478
Anti-MAPT Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Tau ( phospho S199 )

WB: Use at a dilution of 1/1000. Detects a band of approximately 60 kDa. Can be blocked with GTX25241 Tau peptide - phospho S199 (phospho and non-phospho pair) . Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: MAPT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR