Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-STX1A Mouse Monoclonal Antibody [clone: 4H256]

Supplier: Genetex
GTX18010
Syntaxin 1, also known as HPC1, is a 35 kDa integral membrane protein which along with SNAP25, and VAMP/synaptobrevin plays a role in vesicular trafficking and membrane fusion. Two Syntaxin 1 isoforms have been identified, Syntaxin 1A which is localized to nerve terminals of sensory neurons and nerve fibers reaching small blood vessels, and Syntaxin 1B which is localized to motor end plates and muscle spindles. The SNARE (soluble N-ethylmaleimide sensitive fusion protein [NSF] attachment protein [SNAPs] receptors)hypothesis of membrane fusion proposes that Syntaxin 1A and SNAP25 (target membrane SNAREs, t-SNAREs) and VAMP/synaptobrevin (vesicular SNAREs, vSNAREs) bind together to form a tripartite structure that along with soluble cytosolic proteins allows for close membrane apposition of donor and target membranes thereby facilitating membrane fusion. The interaction of Syntaxin 1A with vSNAREs is thought to be negatively regulated by the binding of Munc18 to Syntaxin 1A and this interaction is controlled by Cdk5 phosphorylation of Munc18. Syntaxin 1A can be phosphorylated by casein kinase II and phosphorylation of Syntaxin enhances its interaction with Synaptotagmin.

WB: Use at a concentration of 0.1 μg/ml. Detects a protein doublet at ~38 kDa (predicted molecular weight: 38 kDa). No reactivity was seen with a 35 kDa protein in mouse liver extracts. Some reactivity with higher molecular weight bands of unknown identity can be observed. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: STX1A
Clonality: Monoclonal
Clone: 4H256
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Pig, Rat, Cat, Hamster, Xenopus laevis, Bovine




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
50 μg GTX18010 89360-368 Each (50µG) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol STX1A
Antigen Name Syntaxin 1A
Reactivity Cat
Conjugation Unconjugated
Clonality Monoclonal
Antibody Type Primary
Clone 4H256
Host Mouse
Isotype IgG1
Western Blot Yes
Antigen Synonyms neuron-specific antigen HPC-1, STX1, P35-1, HPC-1, SYN1A, syntaxin-1A
Gene ID 6804
Concentration 1 mg/ml
Immunogen Synaptic vesicle-containing fractions that were immunoprecipitated from human brain homogenates using anti-human Synaptophysin monoclonal antibodies.
Purification Protein G purified
Storage Buffer Preservative: None. Constituents: 50% Glycerol, PBS, 0.1mM PMSF
Storage Temperature Keep as concentrated solution. Store at 4ºC short term. For extended storage aliquot and store at -20ºC or below. Avoid freeze-thaw cycles.
Tested Applications WB

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty