Anti-SNCAIP Goat Polyclonal Antibody

Supplier: GeneTex
GTX11106
89356-536EA 603.88 USD
89356-536
Anti-SNCAIP Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to Synphilin 1

Recommended Starting Dilutions: ELISA: 1/5000 - 1/10000. Immunohistology - Paraffin: Max Dilution 1/400. Western Blotting: Max Dilution 1/2000. Not tested in other applications. Optimal dilution / concentration should be determined by the

Type: Primary
Antigen: SNCAIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR