Anti-SULT1A1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX113560
89357-618EA 582.56 USD
89357-618
Anti-SULT1A1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to SULT1A1 (sulfotransferase family, cytosolic, 1A, phenol-preferring, member 1)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000, IP: 1:100-1:500. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: SULT1A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR