Anti-STAT3 Goat Polyclonal Antibody

Supplier: GeneTex
GTX25073
89361-578EA 607.87 USD
89361-578
Anti-STAT3 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to STAT3 (isoform 1/2)

Approx. 90kDa band observed in lysates of NIH3T3 (calculated MW of 88.0kDa according to NP_644805.1 and of 88.1 according to NP_003141.2). Recommended concentration: 1-3µg/ml. Stains mouse thymic medulla and not the cortex. Recommended overnight staining at 10ug/ml, see picture. Data kindly provided by Dr. Marella de Bruijn, MRC Mol. Haematol. Unit, WIMM, JR Hospital, Oxford. This antibody is expected to recognise isoforms 1 and 2 (as represented by NP_644805.1 and NP_003141.2 respectively)

Type: Primary
Antigen: STAT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR