Anti-SRP1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX112284
89357-040EA 582.76 USD
89357-040
Anti-SRP1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to SRP1 (karyopherin alpha 1 (importin alpha 5))

Suggested starting dilutions are as follows: WB: 1:1000-1:10000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: SRP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR