Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SRC Mouse Monoclonal Antibody

Supplier: Genetex

GTX16885
89351-274EA 377.22 USD InStock
89351-274
Anti-SRC Mouse Monoclonal Antibody
Antibodies
May appear as a doublet due to phosphorylation. May be used to precipitate active Src which can then be used in a kinase reaction.

For IP: Use at an assay dependent dilution. For WB: Use at a concentration of 2 -3 µg/ml. Predicted molecular weight: 59.7 kDa. GTX16885 may appear as a doublet on WB due to phosphorylation. This antibody can be used to precipitate active src kinase. Not suitable for IF, IHC-Fr or IHC-P. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: SRC
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty