Anti-SOX1 Rabbit Monoclonal Antibody [clone: EPR4766]

Supplier: GeneTex

GTX62974
89364-616EA 407.81 USD
89364-616
Anti-SOX1 Rabbit Monoclonal Antibody [clone: EPR4766]
Antibodies
Rabbit monoclonal antibody [EPR4766] to SOX1

Type: Primary
Antigen: SOX1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR