Anti-SOD1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX21222
89360-984EA 766.72 USD
89360-984
Anti-SOD1 Rabbit Polyclonal Antibody
Antibodies
Destroys radicals which are normally produced within the cells and which are toxic to biological systems.

Suitable for immunoblotting (western or dot blot), and ELISA. Recommended Dilution(s): 1:1,000 to 1:4,000 for ELISA. Optimal dilutions should be determined by the researcher.

Type: Primary
Antigen: SOD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR