Anti-SMRT Mouse Polyclonal Antibody

Supplier: GeneTex
GTX70245
89364-808EA 580.2 USD
89364-808
Anti-SMRT Mouse Polyclonal Antibody
Antibodies
Mouse polyclonal antiserum to human SMRT

Type: Primary
Antigen: SMRT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR