Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SMARCA2 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX12165
Chromatin, the physiological packaging structure of histone proteins and DNA, is considered a key element in regulating gene expression. Several complexes involved in transcriptional regulation function by either modifying histones or altering chromatin structure. Postranslational modifications of histones, such as acetylation, phosphorylation and methylation, contribute to the regulation of transcription. The ATP-dependent chromatin-remodeling complexes alter chromatin structure by using the energy of ATP hydrolysis to locally disrupt the association of histones with DNA, displacing the nucleosomes from promoter and enhancer regions, and therefore allowing transcription initiation. Chromatin remodeling complexes have been purified from a variety of organisms, and most cell types contain more than one type of complex. These complexes contain structurally related catalytic subunits, but differ in the way in which they manipulate chromatin. Three families of complexes have been described the SWI/SNF family, ISWI family, and Mi-2 family. The SWI/SNF family of ATP-dependent remodeling complexes was identified in yeast, drosophila, and human. It causes nucleosomes to change structure and/or position in order to allow transcriptional activators to gain access to their target sites. In humans, two conserved ATPase subunits have been identified as hBrm (also designated hSNF2alpha) and Brg1 (also designated as SNF2beta). Brg1 (1,613 amino acids) and hBrm (1,586 amino acids) share approximately 52% identity. Components of the hSWI/SNF complexes have been implicated in a range of cellular events including gene activation, regulation of cell growth, and development. Brg1 and hBrm enhances transcriptional activation by glucocorticoid receptors. Apparently, Brg1 and Brm complexes direct distinct cellular processes by recruitment to specific promoters through protein-protein interactions that are unique to each ATPase. The remodeling complexes were traditionally associated with transcriptional activation. However, SWI/SNF has been found associated with repressor complexes, such as HDAC (histone deacetylase) and Rb (retinoblastoma) in a complex that leads to cell cycle arrest, suggesting that they are associated with transcriptional repression. Antibodies reacting specifically with hBrm/SNF2alpha may be used for studying the effects of chromatin remodeling on gene expression.

IF: Use at a concentration of 1 μg/ml. WB: Use at a concentration of 0.15 μg/ml. Predicted molecular weight: 230 kDa. Staining of SMARCA2 is specifically inhibited by the immunizing peptide. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: SMARCA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Fish




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μl GTX12165 89359-216 Each (100µl) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol SMARCA2
Antigen Name SWI/SNF Related Matrix Associated Actin Dependent Regulator of Chromatin subfamily A member 2
Reactivity Fish
Conjugation Unconjugated
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
Western Blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Antigen Synonyms SNF2, BRM, hBRM, BAF190, SWI2, FLJ36757, SNF2L2, SNF2LA, MGC74511, hSNF2a, Sth1p
Gene ID 6595
Concentration 1.3 mg/ml
Immunogen Synthetic peptide: KEDVEKAKKRRGRPPAE, corresponding to amino acids 1354-1370 of SMARCA2 conjugated to maleimide-activated KLH via an N-terminal added cysteine.
Purification Immunogen affinity purified
Storage Buffer Preservative: 15mM Sodium Azide; Constituents: 1% BSA, 0.01M PBS. pH 7.4
Storage Temperature Keep as concentrated solution. Store at 4ºC short term. For extended storage aliquot and store at -20ºC or below. Avoid freeze-thaw cycles.
Tested Applications ICC/IF, WB

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty