Anti-SMAD2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX38629
89363-054EA 607.22 USD
89363-054
Anti-SMAD2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to SMAD2 (phospho Ser467)

WB: 1:500-1:1000

Type: Primary
Antigen: SMAD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR