Anti-SIRT3 Chicken Polyclonal Antibody

Supplier: GeneTex
GTX85105
89366-814EA 601.4 USD
89366-814
Anti-SIRT3 Chicken Polyclonal Antibody
Antibodies
Chicken polyclonal antibody to SIRT3

Type: Primary
Antigen: SIRT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR