Anti-SIAH1 Goat Polyclonal Antibody

Supplier: GeneTex
GTX22237
89361-198EA 611.09 USD
89361-198
Anti-SIAH1 Goat Polyclonal Antibody
Antibodies
This gene encodes a protein that is a member of the seven in absentia homolog (SIAH) family. The protein is an E3 ligase and is involved in ubiquitination and proteasome-mediated degradation of specific proteins. The activity of this ubiquitin ligase has been implicated in the development of certain forms of Parkinson's disease, the regulation of the cellular response to hypoxia and induction of apoptosis. Alternative splicing results in several additional transcript variants, some encoding different isoforms and others that have not been fully characterized.

Western Blot - Approx 30kDa and 45kDa bands seen in U937 Human Brain lysate. Recommended for use at 2.0-4.0?g/ml.

Type: Primary
Antigen: SIAH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR