Anti-SHANK1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX107701
89355-072EA 557.66 USD
89355-072
Anti-SHANK1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to SHANK1 (SH3 and multiple ankyrin repeat domains 1)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000, IHC-Fr: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: SHANK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR