Anti-SHANK1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX30444
89362-916EA 600.29 USD
89362-916
Anti-SHANK1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to SHANK1

For WB: Use at a dilution of 1:500-1:1000 - Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: SHANK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR