Anti-SETMAR Goat Polyclonal Antibody

Supplier: GeneTex
GTX23823
89361-390EA 611.3 USD
89361-390
Anti-SETMAR Goat Polyclonal Antibody
Antibodies
Goat polyclonal to SETMAR

Western Blot - Approx 70 - 75 kDa band seen in Daudi lysate. Recommended for use at 1.0 - 3.0?g/ml. Not tested in other applications. Optimal working dilutions / concentrations should be determined by the end user.

Type: Primary
Antigen: SETMAR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR