Anti-SEC61G Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX101782
89349-258EA 571.58 USD
89349-258
Anti-SEC61G Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Sec61 gamma subunit (Sec61 gamma subunit)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: SEC61G
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR