Anti-SART1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX111158
89356-600EA 582.76 USD
89356-600
Anti-SART1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to SART1 (squamous cell carcinoma antigen recognized by T cells)

Suggested starting dilutions are as follows: WB: 1:1000-1:10000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: SART1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR