Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SARM1 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX17812
89360-332EA 359.06 USD InStock
89360-332
Anti-SARM1 Rabbit Polyclonal Antibody
Antibodies
Involved in innate immnune response. Acts as a negative regulator of TICAM1/TRIF-dependent Toll-like receptor signaling by inhibiting induction of TLR3- and TLR4-dependent genes. Specifically blocks TICAM1/TRIF-dependent transcription-factor activation and gene induction, without affecting the MYD88-dependent pathway or non-TLR signaling. Negative regulator of NF-kappa-B and IRF activation.

WB: Use at a concentration of 0.5 - 2 μg/ml. Detects a band of approximately 75 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: SARM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty