Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Ig Mouse Monoclonal Antibody [clone: 5F12]

Supplier: Genetex

GTX75921
89367-932EA 359.24 USD InStock
89367-932
Anti-Ig Mouse Monoclonal Antibody [clone: 5F12]
Antibodies
Mouse monoclonal antibody [5F12] to Salmonid immunoglobulin

Recommended Starting Dilutions: ELISA: 1/250 - 1/1000 Western Blotting: 1/250 - 1/1000 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: Ig
Clonality: Monoclonal
Clone: 5F12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Fish

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty