Anti-Salmonella A Mouse Monoclonal Antibody [clone: 513]

Supplier: GeneTex
GTX40949
89363-378EA 609.32 USD
89363-378
Anti-Salmonella A Mouse Monoclonal Antibody [clone: 513]
Antibodies
Salmonella is a rod shaped, motile bacterium, nonsporeforming and Gram negative. There is a widespread occurrence in animals, especially in poultry and swine. Environmental sources of the organism include water, soil, insects, factory surfaces, kitchen surfaces, animal feces, raw meats, raw poultry, and raw seafoods, to name only a few.

Type: Primary
Antigen: Salmonella A
Clonality: Monoclonal
Clone: 513
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR