Anti-S100 Mouse Monoclonal Antibody [clone: SH-B1]

Supplier: GeneTex
GTX73714
89353-260EA 592.68 USD
89353-260
Anti-S100 Mouse Monoclonal Antibody [clone: SH-B1]
Antibodies
This antibody is specific against an epitope located on the b chain (i.e. in S-100a and S-100b) but not on the a chain of S- 100 (i.e. in S-100a and S-100ao). This antibody cross reacts with human, porcine, rabbit, cat and rat tissues. This antibody can be used to localize S-100a and S-100b in various tissue sections.

Type: Primary
Antigen: S100
Clonality: Monoclonal
Clone: SH-B1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR