Anti-S100 Mouse Monoclonal Antibody [clone: 3B10]

Supplier: GeneTex
GTX28334
89352-458EA 726.31 USD
89352-458
Anti-S100 Mouse Monoclonal Antibody [clone: 3B10]
Antibodies
Mouse monoclonal [3B10] to S100 - Astrocyte Marker

ELISA: Use at an assay dependent dilution. IA: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Can be paired for ELISA with GTX10203 Mouse monoclonal [6G1] to S100 . Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: S100
Clonality: Monoclonal
Clone: 3B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR