Anti-S. Pneumoniae Mouse Monoclonal Antibody

Supplier: GeneTex
GTX80794
89366-696EA 395.26 USD
89366-696
Anti-S. Pneumoniae Mouse Monoclonal Antibody
There are more than 85 antigenic types of S. pneumoniae, which are determined by capsule antigens. Altough S. pneumoniae is part of the normal respiratory flora, invasion commonly results in pneumonia as well as meningitis. S. pneumoniae is protected from phagocytosis by its polysaccharide capsule which is also a prominent virulence factor. Infections can usually be treated with antibiotics, although resistant strains are emerging.

Type: Primary
Antigen: S. Pneumoniae
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR