Anti-RNASEA Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX26610
89361-958EA 975.29 USD
89361-958
Anti-RNASEA Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Ribonuclease A

immunoblotting (western or dot blot), ELISA, immunoprecipitation and most immunological methods requiring high titer and specificity A working dilution of 1:5,000 to 1:22,000 of the reconstitution concentration is suggested

Type: Primary
Antigen: RNASEA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR