Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RIT2 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX13090
The Ras super-family comprises a group of structurally related low molecular weight (20-30 kDa) proteins, which bind GTP or GDP and exhibit a low intrinsic GTPase activity. They are involved in signal transduction and the regulation of a variety of cellular processes, including cell growth, transformation, differentiation and morphogenesis, nucleocytoplasmic transport, and apoptosis. Ras proteins promote cancer by disrupting the normal controls of cell proliferation and differentiation. All Ras subgroups (Ras, Rho, Rab, Ran, and ADP ribosylation factor) contain five highly conserved domains (G1-G5) and act as molecular switches by alternating between an active GTP-bound form and an inactive GDP-bound form. Rit2 (Ras-like protein in neurons) and Rit1 (Ras-like protein in tissues) share 50% homology to Ras. Both contain the five highly conserved domains (G1-G5), bind GTP in vitro, and are membrane-associated. However, both lack a CAAX box and their mechanism of membrane association is distinct from the typical Ras protein. Rit is a non-lipid-modified Ras-related protein, which lacks any known recognition signal for C-terminal lipidation. Rit1 and Rit2 share a unique effector domain (DPTIEDAYK) that is 100% conserved between murine and human proteins. Rit1 is expressed ubiquitously, but Rit2 is expressed only in subsets of neurons and may be involved in calcium-mediated signaling within neurons. Rit uses novel effector pathways to regulate proliferation and transformation. It has been suggested that Rit1 and Rit2 play important role in the regulation of signaling pathways, distinct from those controlled by Ras. Ras and associated pathways have also been implicated in the development and maintenance of themammalian retina.

WB: Use at a concentration of 5 - 10 μg/ml. Detects a band of approximately 25 kDa (predicted molecular weight: 26 kDa). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: RIT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
25μg GTX13090 89359-584 Each (25µG) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol RIT2
Antigen Name Ras-like without CAAX 2
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
Western Blot Yes
Antigen Synonyms RIBA, ROC2, RIN
Gene ID 6014
Concentration 1 mg/ml
Immunogen Recombinant His-tagged full length protein (Human).
Purification IgG fraction
Storage Buffer Preservative: 0.08% Sodium Azide; Constituents: PBS. pH 7.3
Tested Applications WB

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty