Anti-RIF1 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX85728
89366-892EA 496.75 USD
89366-892
Anti-RIF1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to RIF1

WB (1:500 - 1:3000), ELISA (1:5000 - 1:40000), IF, IHC-P (5 µg/ml)

Type: Primary
Antigen: RIF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chimpanzee
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR