Anti-RICK Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX28428
89362-758EA 660.6 USD
89362-758
Anti-RICK Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to RICK

For WB: Use at a dilution of 1:900 - 1:1,100. Not tested in other applications. Optimal concentrations/dilutions should be determined by the researcher.

Type: Primary
Antigen: RICK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR