Anti-RHOA Rabbit Polyclonal Antibody

GTX101066
89348-798EA 376.43 USD
89348-798
Anti-RHOA Rabbit Polyclonal Antibody
Rabbit Polyclonal antibody to RhoA (ras homolog gene family, member A)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: RHOA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR