Anti-RGS7 Chicken Polyclonal Antibody

Supplier: GeneTex
GTX14266
89367-298EA 663.11 USD
89367-298
Anti-RGS7 Chicken Polyclonal Antibody
Antibodies
RGS7 inhibits signal transduction by increasing the GTPase activity of G protein alpha subunits thereby driving them into their inactive GDP bound form. Activity on G(o) alpha is specifically enhanced by the RGS6/GNG5 dimer. RGS7 may play a role in synaptic vesicle exocytosis and may play an important role in the rapid regulation of neuronal excitability and the cellular responses to short lived stimulations.

For WB: Use at an assay dependent dilution. Predicted molecular weight: 58 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: RGS7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR