Anti-RGS14 Goat Polyclonal Antibody

Supplier: GeneTex
GTX15720
89360-128EA 581.5 USD
89360-128
Anti-RGS14 Goat Polyclonal Antibody
Antibodies
RGS14 inhibits signal transduction by increasing the GTPase activity of G protein alpha subunits thereby driving them into their inactive GDP bound form. Contains 1 GoLoco domain, 2 Ras binding (RBD) domains, 1 RGS domain.

Approx 65-70kDa band observed in Jurkat lysates (predicted MW of 65kDa according to NP_006471). Recommended for use at 0.5-2µg/ml.

Type: Primary
Antigen: RGS14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR