Anti-RGS10 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX77730
89366-268EA 593.27 USD
89366-268
Anti-RGS10 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to RGS10

For WB: Use at a concentration of 1 µg/ml. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: RGS10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR