Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Respiratory Syncytial Virus Mouse Monoclonal Antibody [clone: B023]

Supplier: Genetex

GTX41184
89363-444EA 325.28 USD InStock
89363-444
Anti-Respiratory Syncytial Virus Mouse Monoclonal Antibody [clone: B023]
Antibodies
Respiratory syncytial virus (RSV) is a major cause of respiratory illness in young children. RSV infection produces a variety of signs and symptoms involving different areas of the respiratory tract, from the nose to the lungs. RSV is a negative sense, enveloped RNA virus. The virion is variable in shape and size with average diameter of between 120 and 300 nm. The 63 kD RSV fusion protein of the RSS 2 strain (subtype A) directs fusion of viral and cellular membranes, results in viral penetration, and can direct fusion of infected cells with adjoining cells, resulting in the formation of syncytia or multi nucleated giant cells.

Type: Primary
Antigen: Respiratory Syncytial Virus
Clonality: Monoclonal
Clone: [B023]
Conjugation: No
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Cow

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty