Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti- Mouse Monoclonal Antibody [clone: 8B10]

Supplier: Genetex

GTX10016
89350-572EA 325.28 USD InStock
89350-572
Anti- Mouse Monoclonal Antibody [clone: 8B10]
Antibodies
Mouse monoclonal [8B10] to Respiratory Syncytial Virus

ELISA: Use at an assay dependent dilution. IF: Use at an assay dependent dilution. Detects RSV in infected cell cultures and clinical material from patients. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen:
Clonality: Monoclonal
Clone: 8B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Respiratory syncytial virus

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty