Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Respiratory Syncytial Virus Rabbit Polyclonal Antibody

Supplier: Genetex

GTX12253
89359-306EA 325.28 USD InStock
89359-306
Anti-Respiratory Syncytial Virus Rabbit Polyclonal Antibody
Antibodies
The 63 kD Respiratory Syncytial Virus (RSV) fusion protein of the RSS-2 strain (subtype A) directs fusion of viral and cellular membranes, resulting in viral penetration, and can direct fusion of infected cells with adjoining cells, resulting in the formation of syncytia.

Type: Primary
Antigen: Respiratory Syncytial Virus
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty