Anti-RECA1 Mouse Monoclonal Antibody

Supplier: GeneTex
GTX76652
89366-172EA 609.75 USD
89366-172
Anti-RECA1 Mouse Monoclonal Antibody
Antibodies
Recognises RECA-1, a cell surface antigen which is expressed by all rat endothelial cells.

Type: Primary
Antigen: RECA1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR