Anti-RAPSN Mouse Monoclonal Antibody [clone: 1234]

Supplier: GeneTex
GTX11423
89357-956EA 640.44 USD
89357-956
Anti-RAPSN Mouse Monoclonal Antibody [clone: 1234]
Antibodies
Nicotinic acetylcholine receptors (AChRs) are organized into high-density postsynaptic clusters at the motor endplate that are critical for efficient synaptic transmission. The organization of AChR’s is carried out by the 43 kDa cytoplasmic protein rapsyn. Studies have indicated that mutations which impair the activity of rapsyn can lead to the formation of congenital myasthenic syndrome (CMS). CMS is characterized by poor development of the postsynaptic region.

Recommended Starting Dilutions:ICC/IF: 2 ug/mlIHC: 1:200WB: 1:1000Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: RAPSN
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Amphibian
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR