Anti-RAD9 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX30297
89367-668EA 612.8 USD
89367-668
Anti-RAD9 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal Antibody to Rad9

Type: Primary
Antigen: RAD9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR