Anti-IgG Goat Polyclonal Antibody

Supplier: GeneTex

GTX77061
89353-466EA 122.65 USD
89353-466
Anti-IgG Goat Polyclonal Antibody
Antibodies
Goat anti-Rabbit IgG Polyclonal antibody

Works in most applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR