Anti-RAB2 Goat Polyclonal Antibody

GTX20794
89360-906EA 395.68 USD
89360-906
Anti-RAB2 Goat Polyclonal Antibody
RAB1 is a member of the RAS oncogene family required for vesicle transport from the ER to the Golgi complex.

Western Blot - Approx 23kDa and 18kDa bands seen in HeLa cell lysates. Recommended for use at 1-3?g/ml.

Type: Primary
Antigen: RAB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR