Anti-PUMILIO 1 Goat Polyclonal Antibody

GTX23717
89352-042EA 407.1 USD
89352-042
Anti-PUMILIO 1 Goat Polyclonal Antibody
Goat polyclonal to Pumilio 1

For IP: Use at a concentration of 1-5 µg/mg lysate. For WB: Use at a dilution of 1:10,000-1:50,000. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: PUMILIO 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Drosophila
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR