Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PTPRT Chicken Polyclonal Antibody

Supplier: Genetex

GTX82152
89368-058EA 457.76 USD InStock
89368-058
Anti-PTPRT Chicken Polyclonal Antibody
Antibodies
Chicken polyclonal to Receptor-type tyrosine-protein phosphatase T

WB (only tested on fusion protein): Use at a dilution of 1:2000. Colorimetric method for signal development.

Type: Primary
Antigen: PTPRT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty