Anti-PTCH2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX70350
89364-930EA 574.44 USD
89364-930
Anti-PTCH2 Rabbit Polyclonal Antibody
Antibodies
Mutations of the human Patched tumor suppressor gene (PTCH) have been identified in individuals with the nevoid basal cell carcinoma syndrome (NBCCS) as well as in sporadic basal cell carcinomas and medulloblastomas. PTCH2 has a frameshift mutation resulting in a truncated PTCH2 protein.

Type: Primary
Antigen: PTCH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR