Anti-PSR Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX10526
89354-174EA 611.61 USD
89354-174
Anti-PSR Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to PSR

IP: Use at an assay dependent dilution. WB: Use at a dilution of 1/1000 using whole extracts of human HeLa or A549 cells. Detects a band of approximately 70 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PSR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR