Anti-GINS2 Rabbit Polyclonal Antibody

GTX106840
89354-800EA 176.03 USD
89354-800
Anti-GINS2 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to PSF2 (GINS complex subunit 2 (Psf2 homolog))

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: GINS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR